Gene Transcripts
- Gene Symbol
- Gm7854
- Entrez Gene ID
- 665934
- Gene Name
- predicted gene 7854
- Gene Aliases
- EG665934, ENSMUSG00000054483
- Chromosome Location
- Chr. 5: 43151686 - 43235354 on Build GRCm38
- UniGene ID
- Mm.381491
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028417.1 |
|
5 |
738 |
|
GenBank mRNA |
AK041797.1 |
- |
5 |
739 |
|
AK138349.1 |
- |
3 |
184 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB232433.2 |
- |
4 |
374 |
|
Chr. 5 - 43152047 - 43191408 [-] |
View Full Product Details