Gene Transcripts
- Gene Symbol
- 4930579K19Rik
- Entrez Gene ID
- 75881
- Gene Name
- RIKEN cDNA 4930579K19 gene
- Gene Aliases
- Chromosome Location
- Chr. 9: 98562460 - 98563611 on Build GRCm38
- UniGene ID
- Mm.483402
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029444.1 |
|
1 |
1012 |
|
GenBank mRNA |
AK016329.1 |
- |
1 |
984 |
|
View Full Product Details