Gene Transcripts
- Gene Symbol
- 4933409K07Rik
- Entrez Gene ID
- 108816
- Gene Name
- RIKEN cDNA 4933409K07 gene
- Gene Aliases
- AV071682, b1402
- Chromosome Location
- Chr. 4|NT_187053: 120552 - 144386 on Build GRCm38
- UniGene ID
- Mm.440578
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033123.1 |
|
5 |
4507 |
|
GenBank mRNA |
BC072647.1 |
- |
5 |
4506 |
|
- Gene Symbol
- Slc38a6
- Entrez Gene ID
- 625098
- Gene Name
- solute carrier family 38, member 6
- Gene Aliases
- AW322671, EG625098
- Chromosome Location
- Chr. 12: 73286794 - 73361998 on Build GRCm38
- UniGene ID
- Mm.32075
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054341.1 |
- |
5 |
1494 |
|
AK140045.1 |
- |
6 |
1634 |
|
BC048389.1 |
- |
5 |
1450 |
|
View Full Product Details