Gene Transcripts
- Gene Symbol
- 1700049L16Rik
- Entrez Gene ID
- 108950
- Gene Name
- hematological and neurological expressed 1-like pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 10: 71979885 - 71980690 on Build GRCm38
- UniGene ID
- Mm.212793
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003644.1 |
|
1 |
485 |
|
GenBank mRNA |
AK075762.1 |
- |
1 |
490 |
|
BC048504.1 |
- |
1 |
485 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV045951.3 |
- |
1 |
682 |
|
Chr. 10 - 71979886 - 71980698 [-] |
View Full Product Details