Gene Transcripts
- Gene Symbol
- 5031426D15Rik
- Entrez Gene ID
- 68144
- Gene Name
- RIKEN cDNA 5031426D15 gene
- Gene Aliases
- Chromosome Location
- Chr. 2: 6922702 - 6928722 on Build GRCm38
- UniGene ID
- Mm.483289
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027890.1 |
|
1 |
221 |
|
GenBank mRNA |
AK019879.1 |
- |
1 |
221 |
|
View Full Product Details