Gene Transcripts
- Gene Symbol
- Gm13315
- Entrez Gene ID
- 625342
- Gene Name
- lactate dehydrogenase A pseudogene
- Gene Aliases
- OTTMUSG00000011384
- Chromosome Location
- Chr. 2: 14720642 - 14722622 on Build GRCm38
- UniGene ID
- Mm.484784
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028497.1 |
|
1 |
1131 |
|
View Full Product Details