Gene Transcripts
- Gene Symbol
- Gm4566
- Entrez Gene ID
- 100043644
- Gene Name
- ubiquitin specific peptidase 16 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 17: 71333201 - 71341804 on Build GRCm38
- UniGene ID
- Mm.362377
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028023.1 |
|
3 |
484 |
|
GenBank mRNA |
CO802078.1 |
- |
3 |
486 |
|
DV658461.1 |
- |
2 |
606 |
|
View Full Product Details