Gene Transcripts
- Gene Symbol
- Mx2
- Entrez Gene ID
- 17858
- Gene Name
- MX dynamin-like GTPase 2
- Gene Aliases
- AI528743, Mx-2
- Chromosome Location
- Chr. 16: 97536081 - 97560901 on Build GRCm38
- UniGene ID
- Mm.14157
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI150753.1 |
- |
1, 2 |
124 |
|
Chr. 16 - 97551396 - 97560428 [+] |
GenBankmRNA |
BI149262.1 |
- |
3, 4 |
274 |
|
Chr. 16 - 97547457 - 97558673 [+] |
GenBankmRNA |
CA544052.1 |
- |
3, 4 |
594 |
|
Chr. 16 - 97550387 - 97551970 [+] |
View Full Product Details