Gene Transcripts
- Gene Symbol
- PAXIP1-AS2
- Entrez Gene ID
- 100132707
- Gene Name
- PAXIP1 antisense RNA 2
- Gene Aliases
- PAXIP1OS
- Chromosome Location
- Chr. 7: 154928517 - 154949490 on Build GRCh38
- UniGene ID
- Hs.586358
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX356348.2 |
- |
3 |
734 |
|
Chr. 7 - 154928517 - 154944683 [+] |
GenBankmRNA |
BX377778.2 |
- |
3 |
734 |
|
Chr. 7 - 154928517 - 154944724 [+] |
View Full Product Details