Gene Transcripts

Gene Symbol
LINC00634
Entrez Gene ID
339674
Gene Name
long intergenic non-protein coding RNA 634
Gene Aliases
BK250D10.8
Chromosome Location
Chr. 22: 41952187 - 41958942 on Build GRCh38
UniGene ID
Hs.517656
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024355.1 2 910
GenBank mRNA BC040153.1 - 2 799
BC068568.1 - 2 910
CR456401.1 - 1 155

View Full Product Details