Gene Transcripts
- Gene Symbol
- LINC00111
- Entrez Gene ID
- 54090
- Gene Name
- long intergenic non-protein coding RNA 111
- Gene Aliases
- C21orf21, NCRNA00111
- Chromosome Location
- Chr. 21: 41679302 - 41697336 on Build GRCh38
- UniGene ID
- Hs.570442
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024367.1 |
|
1 |
59 |
|
GenBank mRNA |
BX099637.1 |
- |
1 |
195 |
|
View Full Product Details