Gene Transcripts
- Gene Symbol
- FAM193B
- Entrez Gene ID
- 54540
- Gene Name
- family with sequence similarity 193 member B
- Gene Aliases
- IRIZIO
- Chromosome Location
- Chr. 5: 177519789 - 177554585 on Build GRCh38
- UniGene ID
- Hs.484289
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI254367.1 |
- |
3 |
626 |
|
Chr. 5 - 177531337 - 177536544 [-] |
GenBankmRNA |
DB119531.1 |
- |
2 |
321 |
|
Chr. 5 - 177531324 - 177532526 [-] |
View Full Product Details