Gene Transcripts
- Gene Symbol
- MIR210
- Entrez Gene ID
- 406992
- Gene Name
- microRNA 210
- Gene Aliases
- MIRN210, mir-210
- Chromosome Location
- Chr. 11: 568089 - 568198 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029623.1 |
|
1 |
83 |
|
View Full Product Details