Gene Transcripts

Gene Symbol
CECR7
Entrez Gene ID
100130418
Gene Name
cat eye syndrome chromosome region, candidate 7 (non-protein coding)
Gene Aliases
SAHL1
Chromosome Location
Chr. 22: 17036570 - 17058792 on Build GRCh38
UniGene ID
Hs.660640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015352.1 4 2604
GenBank mRNA AK098045.1 - 4 1935
AY026052.1 - 4 2604
BC043198.2 - 4 953

View Full Product Details