Gene Transcripts
- Gene Symbol
- LINC01126
- Entrez Gene ID
- 100129726
- Gene Name
- long intergenic non-protein coding RNA 1126
- Gene Aliases
- Chromosome Location
- Chr. 2: 43227211 - 43228855 on Build GRCh38
- UniGene ID
- Hs.194480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027251.1 |
|
1 |
1476 |
|
GenBank mRNA |
BC032412.1 |
- |
1 |
1346 |
|
View Full Product Details