Gene Transcripts
- Gene Symbol
- TPTEP1
- Entrez Gene ID
- 387590
- Gene Name
- transmembrane phosphatase with tensin homology pseudogene 1
- Gene Aliases
- psiTPTE22
- Chromosome Location
- Chr. 22: 16601911 - 16648830 on Build GRCh38
- UniGene ID
- Hs.474116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001591.1 |
|
8 |
1071 |
|
GenBank mRNA |
AY219889.1 |
- |
8 |
1071 |
|
BC107743.1 |
- |
6 |
613 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA427417.1 |
- |
2 |
193 |
|
Chr. 22 - 16638616 - 16653638 [+] |
GenBankmRNA |
DA281648.1 |
- |
3 |
497 |
|
Chr. 22 - 16601950 - 16648546 [+] |
View Full Product Details