Gene Transcripts
- Gene Symbol
- PP7080
- Entrez Gene ID
- 25845
- Gene Name
- uncharacterized LOC25845
- Gene Aliases
- Chromosome Location
- Chr. 5: 470510 - 472965 on Build GRCh38
- UniGene ID
- Hs.719132
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK023178.1 |
- |
1 |
2371 |
|
Chr. 5 - 470511 - 473063 [-] |
GenBankmRNA |
BC014166.1 |
- |
1 |
835 |
|
Chr. 5 - 469741 - 471527 [-] |
View Full Product Details