Gene Transcripts
- Gene Symbol
- DICER1-AS1
- Entrez Gene ID
- 400242
- Gene Name
- DICER1 antisense RNA 1
- Gene Aliases
- DICER1-AS
- Chromosome Location
- Chr. 14: 95157688 - 95179933 on Build GRCh38
- UniGene ID
- Hs.497573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC040596.1 |
- |
1 |
2591 |
|
View Full Product Details