Gene Transcripts
- Gene Symbol
- MCM3AP-AS1
- Entrez Gene ID
- 114044
- Gene Name
- MCM3AP antisense RNA 1
- Gene Aliases
- C21orf85, MCM3AP-AS, MCM3APAS, MCM3APASB, NCRNA00031
- Chromosome Location
- Chr. 21: 46229231 - 46251701 on Build GRCh38
- UniGene ID
- Hs.655095
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110566.1 |
|
3 |
1017 |
|
NR_110567.1 |
|
2 |
1008 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF426262.1 |
- |
2 |
992 |
|
Chr. 21 - 46229247 - 46243001 [+] |
View Full Product Details