Gene Transcripts
- Gene Symbol
- FLJ45950
- Entrez Gene ID
- 399975
- Gene Name
- FLJ45950 protein
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127847.1 |
- |
2 |
3114 |
|
View Full Product Details