Gene Transcripts
- Gene Symbol
- LOC100128494
- Entrez Gene ID
- 100128494
- Gene Name
- uncharacterized LOC100128494
- Gene Aliases
- Chromosome Location
- Chr. 11: 72014291 - 72020910 on Build GRCh38
- UniGene ID
- Hs.713607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104178.1 |
|
2 |
2831 |
|
GenBank mRNA |
AK126629.1 |
- |
2 |
2832 |
|
View Full Product Details