Gene Transcripts
- Gene Symbol
- AGAP1
- Entrez Gene ID
- 116987
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
- Gene Aliases
- AGAP-1, CENTG2, GGAP1, cnt-g2
- Chromosome Location
- Chr. 2: 235494089 - 236131800 on Build GRCh38
- UniGene ID
- Hs.435039
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR608907.1 |
- |
1 |
1855 |
|
Chr. 2 - 236129677 - 236131772 [+] |
GenBankmRNA |
BX451605.2 |
- |
1 |
481 |
|
Chr. 2 - 236131077 - 236131793 [+] |
GenBankmRNA |
BX096592.1 |
- |
1 |
451 |
|
Chr. 2 - 236131081 - 236131738 [+] |
GenBankmRNA |
EL952667.1 |
- |
1 |
344 |
|
Chr. 2 - 236131191 - 236131725 [+] |
View Full Product Details