Gene Transcripts
- Gene Symbol
- ART3
- Entrez Gene ID
- 419
- Gene Name
- ADP-ribosyltransferase 3
- Gene Aliases
- ARTC3
- Chromosome Location
- Chr. 4: 76011180 - 76112802 on Build GRCh38
- UniGene ID
- Hs.24976
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK129914.1 |
- |
1 |
1465 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX096471.1 |
- |
1 |
140 |
|
Chr. 4 - 76108151 - 76108572 [+] |
View Full Product Details