Gene Transcripts
- Gene Symbol
- NCBP3
- Entrez Gene ID
- 55421
- Gene Name
- nuclear cap binding subunit 3
- Gene Aliases
- C17orf85, ELG, HSA277841
- Chromosome Location
- Chr. 17: 3806748 - 3846251 on Build GRCh38
- UniGene ID
- Hs.120963
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG675548.1 |
- |
1 |
35 |
|
Chr. 17 - 3807416 - 3808194 [-] |
GenBankmRNA |
AA284923.1 |
- |
1 |
323 |
|
Chr. 17 - 3808033 - 3808484 [-] |
View Full Product Details