Gene Transcripts
- Gene Symbol
- FAM234B
- Entrez Gene ID
- 57613
- Gene Name
- family with sequence similarity 234 member B
- Gene Aliases
- KIAA1467
- Chromosome Location
- Chr. 12: 12927680 - 13083449 on Build GRCh38
- UniGene ID
- Hs.132660
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125676.1 |
- |
3 |
539 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA243448.1 |
- |
3 |
545 |
|
Chr. 12 - 13008094 - 13019966 [+] |
View Full Product Details