Gene Transcripts
- Gene Symbol
- RALGAPA1
- Entrez Gene ID
- 253959
- Gene Name
- Ral GTPase activating protein catalytic alpha subunit 1
- Gene Aliases
- GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
- Chromosome Location
- Chr. 14: 35538352 - 35809304 on Build GRCh38
- UniGene ID
- Hs.113150
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR599879.1 |
- |
1 |
463 |
|
Chr. 14 - 35541101 - 35542558 [-] |
View Full Product Details