Gene Transcripts
- Gene Symbol
- KCNK9
- Entrez Gene ID
- 51305
- Gene Name
- potassium two pore domain channel subfamily K member 9
- Gene Aliases
- K2p9.1, KT3.2, TASK-3, TASK3
- Chromosome Location
- Chr. 8: 139600838 - 139703135 on Build GRCh38
- UniGene ID
- Hs.493037
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094401.1 |
- |
1 |
1453 |
|
View Full Product Details