Gene Transcripts
- Gene Symbol
- LOC101927415
- Entrez Gene ID
- 101927415
- Gene Name
- uncharacterized LOC101927415
- Gene Aliases
- Chromosome Location
- Chr. 12: 123581066 - 123584327 on Build GRCh38
- UniGene ID
- Hs.636524
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110049.2 |
|
3 |
1150 |
|
GenBank mRNA |
AK125282.1 |
- |
3 |
1150 |
|
View Full Product Details