Gene Transcripts
- Gene Symbol
- LOC729224
- Entrez Gene ID
- 729224
- Gene Name
- uncharacterized LOC729224
- Gene Aliases
- Chromosome Location
- Chr. 2: 202087555 - 202117058 on Build GRCh38
- UniGene ID
- Hs.623895
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC040292.1 |
- |
1 |
1038 |
|
View Full Product Details