Gene Transcripts
- Gene Symbol
- LOC100422212
- Entrez Gene ID
- 100422212
- Gene Name
- eukaryotic translation initiation factor 3 subunit J pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 1: 163421070 - 163423191 on Build GRCh38
- UniGene ID
- Hs.556898
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104294.1 |
|
1 |
1382 |
|
GenBank mRNA |
BC036607.1 |
- |
1 |
1377 |
|
BC047397.1 |
- |
1 |
1383 |
|
View Full Product Details