Gene Transcripts
- Gene Symbol
- CNTN6
- Entrez Gene ID
- 27255
- Gene Name
- contactin 6
- Gene Aliases
- NB3
- Chromosome Location
- Chr. 3: 978021 - 1403610 on Build GRCh38
- UniGene ID
- Hs.387300
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125798.1 |
- |
1 |
43 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB061357.1 |
- |
1 |
43 |
|
Chr. 3 - 1008141 - 1009787 [+] |
View Full Product Details