Gene Transcripts
- Gene Symbol
- TNRC6C
- Entrez Gene ID
- 57690
- Gene Name
- trinucleotide repeat containing 6C
- Gene Aliases
- Chromosome Location
- Chr. 17: 77974984 - 78108835 on Build GRCh38
- UniGene ID
- Hs.406810
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054995.1 |
- |
1 |
1547 |
|
View Full Product Details