Gene Transcripts
- Gene Symbol
- LOC283737
- Entrez Gene ID
- 283737
- Gene Name
- uncharacterized LOC283737
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.31795
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096897.1 |
- |
1 |
656 |
|
- Gene Symbol
- AGBL1
- Entrez Gene ID
- 123624
- Gene Name
- ATP/GTP binding protein like 1
- Gene Aliases
- CCP4, FECD8
- Chromosome Location
- Chr. 15: 86079635 - 87031476 on Build GRCh38
- UniGene ID
- Hs.31795
- Species
- Homo sapiens
View Full Product Details