Gene Transcripts
- Gene Symbol
- TRAF3IP2-AS1
- Entrez Gene ID
- 643749
- Gene Name
- TRAF3IP2 antisense RNA 1
- Gene Aliases
- C6UAS, C6orf3, NCRNA00248, TRAF3IP2-AS2
- Chromosome Location
- Chr. 6: 111483472 - 111602295 on Build GRCh38
- UniGene ID
- Hs.486228
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034108.1 |
|
4 |
2909 |
|
NR_034109.1 |
|
2 |
2618 |
|
GenBank mRNA |
AF136408.1 |
- |
1 |
387 |
|
View Full Product Details