Gene Transcripts
- Gene Symbol
- SP100
- Entrez Gene ID
- 6672
- Gene Name
- SP100 nuclear antigen
- Gene Aliases
- lysp100b
- Chromosome Location
- Chr. 2: 230416156 - 230545602 on Build GRCh38
- UniGene ID
- Hs.369056
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L79989.1 |
- |
1 |
525 |
|
View Full Product Details