Gene Transcripts
- Gene Symbol
- LINC00458
- Entrez Gene ID
- 100507428
- Gene Name
- long intergenic non-protein coding RNA 458
- Gene Aliases
- LncRNA-ES3
- Chromosome Location
- Chr. 13: 54124324 - 54132871 on Build GRCh38
- UniGene ID
- Hs.351262
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_108063.1 |
|
3 |
793 |
|
GenBank mRNA |
BC017942.1 |
- |
3 |
788 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CX167424.1 |
- |
3 |
742 |
|
Chr. 13 - 54126963 - 54132820 [-] |
View Full Product Details