Gene Transcripts
- Gene Symbol
- 4930432J09Rik
- Entrez Gene ID
- 73877
- Gene Name
- RIKEN cDNA 4930432J09 gene
- Gene Aliases
- Chromosome Location
- Chr. 14: 103977628 - 104027142 on Build GRCm38
- UniGene ID
- Mm.399172
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045953.1 |
|
3 |
966 |
|
GenBank mRNA |
AK015289.1 |
- |
3 |
920 |
|
AK052661.1 |
- |
3 |
967 |
|
View Full Product Details