Gene Transcripts

Gene Symbol
4933432G23Rik
Entrez Gene ID
71196
Gene Name
RIKEN cDNA 4933432G23 gene
Gene Aliases
Chromosome Location
Chr. 9: 118429839 - 118437048 on Build GRCm38
UniGene ID
Mm.73218
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_040716.1 2 257
GenBank mRNA AK017021.1 - 2 261


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY716705.1 - 2 257 Chr. 9 - 118429839 - 118437039 [+]

View Full Product Details