Gene Transcripts
- Gene Symbol
- 2010320O07Rik
- Entrez Gene ID
- 69881
- Gene Name
- RIKEN cDNA 2010320O07 gene
- Gene Aliases
- Chromosome Location
- Chr. 18: 38212112 - 38229163 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008598.1 |
- |
|
164 |
|
View Full Product Details