Gene Transcripts
- Gene Symbol
- 4833415N18Rik
- Entrez Gene ID
- 73761
- Gene Name
- RIKEN cDNA 4833415N18 gene
- Gene Aliases
- Chromosome Location
- Chr. 16: 11779702 - 11781501 on Build GRCm38
- UniGene ID
- Mm.39183
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014704.1 |
- |
2 |
253 |
|
BE653989.1 |
- |
2 |
274 |
|
View Full Product Details