Gene Transcripts
- Gene Symbol
- Helz
- Entrez Gene ID
- 78455
- Gene Name
- helicase with zinc finger domain
- Gene Aliases
- 3110078M01Rik, 9430093I07Rik, 9630002H22Rik, AI851979, DRHC
- Chromosome Location
- Chr. 11: 107546958 - 107693826 on Build GRCm38
- UniGene ID
- Mm.272047
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014258.1 |
- |
1 |
236 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF464744.1 |
- |
1 |
258 |
|
Chr. 11 - 107690641 - 107691022 [+] |
GenBankmRNA |
BG148557.1 |
- |
1 |
198 |
|
Chr. 11 - 107690701 - 107691084 [+] |
View Full Product Details