Gene Transcripts
- Gene Symbol
- Gm11418
- Entrez Gene ID
- 102634628
- Gene Name
- predicted gene 11418
- Gene Aliases
- OTTMUSG00000000855
- Chromosome Location
- Chr. 11: 81586945 - 81600950 on Build GRCm38
- UniGene ID
- Mm.435885
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK132715.1 |
- |
3 |
288 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB614803.1 |
- |
3 |
288 |
|
Chr. 11 - 81587359 - 81593480 [+] |
View Full Product Details