Gene Transcripts
- Gene Symbol
- Gm15417
- Entrez Gene ID
- 545539
- Gene Name
- predicted gene 15417
- Gene Aliases
- ENSMUSG00000074466, OTTMUSG00000021274
- Chromosome Location
- Chr. 3: 89391864 - 89398779 on Build GRCm38
- UniGene ID
- Mm.393389
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040403.1 |
|
2 |
1123 |
|
NR_040404.1 |
|
2 |
584 |
|
GenBank mRNA |
AK136654.1 |
- |
2 |
585 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY721031.1 |
- |
2 |
581 |
|
Chr. 3 - 89394293 - 89398530 [+] |
GenBankmRNA |
CX210895.1 |
- |
1 |
257 |
|
Chr. 3 - 89398182 - 89398777 [+] |
View Full Product Details