Gene Transcripts

Gene Symbol
Gdap6
Entrez Gene ID
14551
Gene Name
ganglioside-induced differentiation-associated-protein 6
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.453134
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA Y17855.1 - 1 729

View Full Product Details