Gene Transcripts
- Gene Symbol
- Fabp7
- Entrez Gene ID
- 12140
- Gene Name
- fatty acid binding protein 7, brain
- Gene Aliases
- B-FABP, BFABP, Blbp, MRG
- Chromosome Location
- Chr. 10: 57784923 - 57788450 on Build GRCm38
- UniGene ID
- Mm.3644
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK163791.1 |
- |
1 |
1810 |
|
View Full Product Details