Gene Transcripts
- Gene Symbol
- Zfp346
- Entrez Gene ID
- 26919
- Gene Name
- zinc finger protein 346
- Gene Aliases
- Jaz, Znf346
- Chromosome Location
- Chr. 13: 55105271 - 55135071 on Build GRCm38
- UniGene ID
- Mm.22795
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK084710.1 |
- |
1 |
2160 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB478760.2 |
- |
1 |
656 |
|
Chr. 13 - 55134504 - 55135170 [+] |
View Full Product Details