Gene Transcripts
- Gene Symbol
- Gm38575
- Entrez Gene ID
- 102641783
- Gene Name
- predicted gene, 38575
- Gene Aliases
- Chromosome Location
- Chr. 10: 85230038 - 85266429 on Build GRCm38
- UniGene ID
- Mm.213804
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038448.1 |
- |
|
191 |
|
AK165297.1 |
- |
|
122 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB632197.1 |
- |
1 |
191 |
|
Chr. 10 - 85231572 - 85266392 [-] |
GenBankmRNA |
BB843340.1 |
- |
1 |
122 |
|
Chr. 10 - 85231783 - 85266322 [-] |
View Full Product Details