Gene Transcripts
- Gene Symbol
- Gm15614
- Entrez Gene ID
- 102640951
- Gene Name
- predicted gene 15614
- Gene Aliases
- OTTMUSG00000024330
- Chromosome Location
- Chr. 5: 32507184 - 32583386 on Build GRCm38
- UniGene ID
- Mm.389614
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136552.1 |
- |
2 |
293 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ054039.1 |
- |
2 |
293 |
|
Chr. 5 - 32508099 - 32583386 [-] |
View Full Product Details