Gene Transcripts

Gene Symbol
Socs4
Entrez Gene ID
67296
Gene Name
suppressor of cytokine signaling 4
Gene Aliases
3110032M18Rik, A730004F22Rik, AI427843, AU015727, AU015859, Socs7
Chromosome Location
Chr. 14: 47277143 - 47291591 on Build GRCm38
UniGene ID
Mm.23957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK080408.1 - 1954

View Full Product Details