Gene Transcripts
- Gene Symbol
- Gm29811
- Entrez Gene ID
- 101243624
- Gene Name
- predicted gene, 29811
- Gene Aliases
- Chromosome Location
- Chr. 3: 132842202 - 132878596 on Build GRCm38
- UniGene ID
- Mm.306256
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_102297.1 |
|
4 |
1385 |
|
NR_102298.1 |
|
4 |
1382 |
|
GenBank mRNA |
AK135894.1 |
- |
4 |
1386 |
|
View Full Product Details